Invitrogen™

Ambion™ RNase I, cloned, 100 U/µL

Catalog number:  AM2295

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® RNase I efficiently cleaves after all four bases of single-stranded RNA, in contrast to RNase A, which only cleaves after C and U residues. RNase I degrades all RNA dinucleotide bonds leaving a 5' hydroxyl and 2', 3' cyclic monophosphate. Supplied in one tube of 25,000 U (100 U/ µL). RNase I will degrade any RNA to a mixture of mononucleotides, dinucleotides, and trinucleotides and does not degrade DNA, although it will bind to DNA. It has a marked preference for single-stranded RNA over double-stranded RNA, which allows it to work well in ribonuclease protection assays. This RNase I is rigorously tested for nonspecific contaminating endonuclease, exonuclease, and protease activity.

Unit Definition:
One unit is the amount of enzyme required to produce 1 µg of acid-soluble material from mouse liver RNA in 30 min at 37°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Enzyme: RNase
Product Line: Ambion™
Product Size: 25,000 units
Shipping Condition: Dry Ice

Contents & storage

Store at –20°C

Documents

Manuals & protocols