This product will be discontinued on September 1, 2016. We recommend you switch to SUPERase• In™ RNase Inhibitor (20 U/µL).

ANTI-RNase (15-30 U/μL)

Catalog number: AM2692

Ambion™  Related applications: In Vitro Transcription | Real Time PCR (qPCR) | Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® ANTI-RNase is a protein ribonuclease inhibitor that noncovalently binds and inactivates neutral pancreatic RNase A–type enzymes. It is supplied in one tube containing 10,000 U (15–30 U⁄μL). No inhibition is seen with RNase 1, T1, T2, H, U1, U2, or CL3. ANTI-RNase is distinct from human placental ribonuclease inhibitor in that it has a more robust interaction with RNase and does not release active RNase in the absence of DTT or other reducing agents. ANTI-RNase may be used in any application where placental ribonuclease inhibitor is used. It is ideal for use in invitro transcription and translation, cDNA synthesis PCR, RT-PCR, RNA isolation procedures, and the preparation of RNase-free antibodies. ANTI-RNase is added to reactions to give a final concentration of 1 U⁄μL. Slight inhibition of ANTI-RNase is measured in the presence of DTT (<=20 mM). ANTI-RNase is thoroughly tested for contaminating RNase, exonuclease, and protease activity.

Unit Definition:


One unit is the amount of protein required to inhibit the activity of 5 ng RNase A by 50%.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 10,000 units
Shipping Condition: Dry Ice

Contents & storage

Store at -20°C. Do not store in a frost–free freezer or at -70°C.

Documents

Manuals & protocols