siPORT™ Amine Transfection Agent

Catalog number: AM4503

Invitrogen™  Related applications: Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A polyamine-based Ambion® siRNA transfection agent that can be used to transfect cells during subculturing-—saving a full day of valuable time—without increasing toxicity. It is supplied in one tube containing 1 mL. A successful siRNA experiment requires the appropriate transfection agent for your cells, and one that is optimized for siRNA delivery. The efficiency with which different transfection reagents deliver nucleic acids can vary dramatically with the type of nucleic acid being transfected (RNA vs. DNA, single-stranded vs. double-stranded) and the cells being used.

Accessory Products:
siPORT™ NeoFX™ Transfection Agent (SKU#s AM4511 and AM4510), a cationic and neutral lipid mixture, is also available. For testing which transfection agent is best with your system, the Silencer® siRNA Transfection II Kit (SKU# AM1631) is recommended. It contains the two different transfection agents along with controls for transfection optimization.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Established Cell Lines, Primary Cells
Sample Type (Specific): Synthetic siRNA
Classification: Animal Origin-Free
Transfection Technique: Lipid-Based Transfection
Serum Compatible: Yes
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Line: Ambion™, siPORT™
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store at 4°C.

Documents

Manuals & protocols

Recommended products