Invitrogen™

siPORT™ NeoFX™ Transfection Agent with manual

Catalog number:  AM4511M

 Related applications: Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Ambion® lipid-based formulation can be used to efficiently transfect adherent cells with siRNA as they are subcultured—without increased cytotoxicity. It is provided in one tube containing 1 mL.

• Versatile—works with a broad range of cell lines
• Reproducible—produces consistent results, lot-to-lot, plate-to-plate, and well-to-well
• Efficient—performs with high siRNA transfection efficiency, allowing use of low siRNA concentrations to minimize nonspecific effects
• Fast—reverse-transfects cells as they are plated, saving a day

Gene silencing experiments often call for critical, but time-consuming optimization experiments. siPORT™ NeoFX Transfection Agent refines siRNA transfection protocols resulting in less optimization. From start to finish, successful gene silencing experiments can be completed in as early as 24 hours. This streamlined protocol can be adapted to a wide range of cells and experimental designs, including high-throughput applications.

Reverse-Transfect Cells as They are Plated, Saving a Day
Just add siPORT™ NeoFX to your diluted siRNA, incubate to form transfection complexes, add the complexes to the culture wells, and overlay with cells. This new streamlined protocol, named "neofection", will save researchers a full day of valuable time when compared to traditional plated transfection procedures. The transfection complexes are active and stable even in the presence of serum, so there is no need to remove or replace media following transfection.

Optimized for Lower siRNA Concentrations and High Reproducibility
Recent reports indicate that siRNA concentrations of 100 nM or higher can lead to nonspecific changes in gene expression (off-target effects) in mammalian cultured cells. Reducing the amount of siRNA used for transfections to 1–20 nM minimizes these nonspecific effects, while still providing extensive silencing of the target gene. siPORT™ NeoFX Transfection Agent efficiently transfects low concentrations of siRNA. Furthermore, this agent delivers high performance among multiple wells for replicate samples and between different lots of the transfection agent.

Compatible with a Broad Range of Cell Lines
The agent has been used successfully to deliver siRNA into many cell types, including MCF-7 (human breast adenocarcinoma), HT-29 (human colorectal adenocarcinoma), HeLa-S3 (human cervix adenocarcinoma), HeLa (human cervix adenocarcinoma), HepG2 (human hepatocellular carcinoma), BJ (human foreskin fibroblast), A549 (human lung carcinoma), UMR106 (rat osteosarcoma), SKOv3 (human ovarian carcinoma), and SKNAS (human bone marrow neuroblastoma).

Accessory Products:
siPORT™ Amine Transfection Agent (SKU#s AM4502 and AM4503), a polyamine agent, is also available. For testing which transfection agent is best with your system, the Silencer® siRNA Transfection II Kit (SKU# AM1631) is recommended. It contains the two different transfection agents along with controls for transfection optimization.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Established Cell Lines, Primary Cells, Stem Cells
Sample Type (Specific): Synthetic siRNA
Transfection Technique: Lipid-Based Transfection
Serum Compatible: Yes
High Throughput Compatibility: High Throughput-Compatible
Product Line: Ambion™, siPORT™
Product Size: 1 mL

Contents & storage

Store at 4°C.

Documents

Manuals & protocols