THE RNA Storage Solution

Catalog number: AM7001

Invitrogen™  Related applications: RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

THE Ambion® RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer. This 1 mM sodium citrate, pH 6.4 +/- 0.2 buffer is provided in one bottle containing 50 mL. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. THE RNA Storage Solution has two features that minimize base hydrolysis of RNA: a low pH, and sodium citrate, which is an efficient chelating agent. THE RNA Storage Solution is compatible with all common RNA applications such as reverse transcription, in vitro transcription, northern analysis, and nuclease protection assays. THE RNA Storage Solution is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: RNA Storage Reagent
Form: Liquid
Product Line: Ambion™
Product Size: 50 mL
Shipping Condition: Room Temperature

Contents & storage

Store at -20°C.
Shipped at ambient temperature.

Documents

Manuals & protocols