RNAsecure™ RNase Inactivation Reagent

Catalog number:  AM7006

Invitrogen™  Related applications: RNA Extraction | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® RNAsecure (patents pending) is a unique non-enzymatic reagent that will irreversibly inactivate RNases in solution. 10 mL supplied.
• No post-treatment autoclaving
• Safe, easy-to-use reagent inactivates RNases in solutions
• Inactivation process can be repeated to protect against newly introduced contaminants
• Compatible with downstream procedures, such as RT-PCR and in vitro transcription
• Inactivate RNases in Tris and other solutions that cannot be treated with DEPC RNase contamination in reagents used for RNA isolation and analysis can contribute to experimental inconsistency and, at worst, can cause experimental failure. Traditionally, DEPC has been used to treat solutions that come into contact with RNA. DEPC treatment is time-consuming, possibly hazardous, and only eliminates RNases present at the time of DEPC treatment. In addition, some solutions (e.g., primary amine containing compounds such as Tris) cannot be treated with DEPC at all. Finally, DEPC has to be inactivated by autoclaving post-treatment to prevent it from interfering with downstream enzymatic reactions. RNAsecure Reagent eliminates these problems. Unlike DEPC, RNAsecure can be used on virtually any solution and does not require post-treatment autoclaving. A unique feature of RNAsecure is that reheating after the initial treatment will reactivate the RNase-destroying agent to eliminate any new contaminants. RNAsecure is supplied as a 25X concentrated stock. It is added to solutions as part of reactions (in vitro transcription, RT-PCR, etc.), prior to the addition of enzymes, and heated to 60°C for 10 min to inactivate RNase.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: RNase Control Reagent
Form: Liquid
Product Line: Ambion™, RNAsecure
Product Size: 10 mL
Purity or Quality Grade: Molecular Biology Grade
Shipping Condition: Dry Ice

Contents & storage

Store RNAsecure™ at -20°C.

Documents

Manuals & protocols