Invitrogen™

RNAsecure™ Resuspension Solution (1 ml Tube)

Catalog number:  AM7010

 Related applications: RNA Extraction | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® RNAsecure Resuspension Solution (patents pending) is a unique non-enzymatic reagent that will irreversibly inactivate RNases in solution. It is supplied at working concentration for direct resuspension of RNA pellets. Ten tubes containing 1 mL each are provided.

• No post-treatment autoclaving
• Safe, easy-to-use reagent inactivates RNases in solution
• Inactivation process can be repeated to protect against newly introduced contaminants
• Compatible with downstream procedures, such as RT-PCR and in vitro transcription

RNAsecure™ Resuspension Solution is specifically meant for resuspending RNA pellets that may contain trace amounts of ribonuclease (e.g., after total RNA isolation). Note: it cannot reverse RNA degradation that has already occurred, but can prevent further RNA degradation. To use, the RNA pellet is resuspended in the RNAsecure Resuspension Solution and heated to 60°C for 10 min. It can be reheated to eliminate any RNase contamination introduced at a later time.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: RNase Control Reagent
Form: Liquid
Product Line: Ambion™, RNAsecure
Product Size: 10 tubes
Purity or Quality Grade: Molecular Biology Grade
Shipping Condition: Dry Ice

Contents & storage

Store pure solution at –20°C.

Documents

Manuals & protocols