Decade™ Markers System (to make radiolabeled RNA markers)

Catalog number: AM7778

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Ambion® Decade™ Marker System produces a set of radiolabeled RNA molecules of 150, 100, 90, 80, 70, 60, 50, 40, 30, 20, and 10 nucleotides. The molecular weight markers are ideally suited for experiments where RNA molecules less than 150 nucleotides in length are being studied. Sufficient reagents are provided for 10 marker preparation reactions. This marker set is particularly useful for research laboratories studying tRNA, snRNA, snoRNA, miRNA, siRNA and other small RNA molecules. The system includes all of the reagents necessary to produce the molecular weight markers except gamma-32P-ATP. The Decade™ Markers derive from a single, gel-purified transcript that is provided. The transcript is 5' end-labeled using the kinase and kinase buffer supplied with the kit. The kinase reaction is then diluted into a cleavage reagent that generates the molecular weight marker set in a five minute, room temperature reaction. Gel loading buffer from the kit is then added and the marker is ready for electrophoresis. The labeling, cleavage, and gel preparation steps can be completed in less than an hour.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: 100 µg⁄ml
Gel Compatibility: Denaturing Polyacrylamide Gels
Product Line: Ambion™, Decade™
Product Size: 10 reactions
Ready to Load: No
Size Range: 10 to 150 bp
Shipping Condition: Dry Ice

Contents & storage

Decade™ Marker RNA should be stored at below -70°C. Polynucleotide Kinase, Kinase Reaction Buffer, Cleavage Reagent, Gel Loading Buffer II are stored at 4°C or -20°C. Nuclease-free water is stored at any temperature.

Documents

Manuals & protocols

Recommended products