MagMAX™ Lysis/Binding Solution Concentrate

Catalog number: AM8500

Ambion™  Related applications: Animal Health | Automated Nucleic Acid Purification | RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® MagMAX™ Lysis⁄Binding Solution Concentrate is a component of all the MagMAX™ Isolation Kits that may be purchased separately. Provided in one 100 mL bottle.

This solution is the same formulation as the lysis buffer used in Cat. Nos. AM1830, AM1835, AM1836, AM1837, AM1839, AM1840, 4462359, and A27828.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Binding Buffer, Lysis Buffer
Product Line: Ambion™, MagMAX™
Product Size: 100 mL
Shipping Condition: Room Temperature

Contents & storage

Store MagMAX™ Lysis⁄Binding Solution Concentrate at room temperature.

Documents

Manuals & protocols

Recommended products