Gel Loading Buffer II (Denaturing PAGE)

Catalog number:  AM8547

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue. Supplied in one 10 mL bottle. All Ambion® Gel Loading Solutions are rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity, and for functionality. These are the same solutions we use in-house and in our kits.
For Research Use Only. Not for use in diagnostic procedures.


Buffer Type: Sample Loading Buffers
Gel Compatibility: Denaturing Polyacrylamide Gels
Label or Dye: Bromophenol Blue, Xylene Cyanol
Product Line: Ambion™
Product Size: 10 mL
Shipping Condition: Room Temperature

Contents & storage

Store at –20°C.


Manuals & protocols

Recommended products