NorthernMax™ One-Hour Transfer Buffer

Catalog number: AM8672

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Ambion® NorthernMax® Transfer Buffer is faster and more efficient than standard, overnight transfer buffer. Transfer is complete in 1.5–2 hr. One bottle containing 1 L is provided. Transfer buffer is not compatible with nitrocellulose membrane. NorthernMax® reagents are the same components included in the NorthernMax® Ultrasensitive Northern Blotting Kits but in larger sizes. These reagents are supplied ready to use and are certified RNase-free. Comprehensive instructions are included.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Ambion™, NorthernMax®
Product Size: 1 L
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products