Acrylamide/Bis 19:1, 40% (w/v) solution

Catalog number: AM9022

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® Acrylamide/Bis 19:1 is a 40% (w/v) solution of acrylamide (38%) and bis-acrylamide (2%) ideal for use in ribonuclease protection assay, sequencing gels, and sizing DNA or RNA fragments. Supplied in one bottle containing 500 mL. The solution is provided in a ready-to-use form, reducing the dust, inhalation, and contact hazard associated with weighing and preparing acrylamide and bis-acrylamide powders and solutions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Ambion™
Product Size: 500 mL
Shipping Condition: Room Temperature

Contents & storage

Store at 4°C.

Documents

Manuals & protocols

Recommended products