RNaseZap™ RNase Decontamination Solution

Catalog number: AM9784

Invitrogen™  Related applications: RNA Extraction | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

RNaseZap® RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap® Solution onto the surface to be decontaminated and rinse it off with RNase-free water. Features of RNaseZap® RNase Decontamination Solution:

• Completely removes RNase contamination from glass and plastic surfaces
• Excels at removing high levels of RNase contamination whereas similar products fail
• Proven effective at removing high concentrations of dried-on RNase A
• Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free

Using RNaseZap® Solution
Working with RNA requires that special measures be taken to ensure an RNase-free environment. Even trace quantities of RNase can lead to lower yields from in vitro transcription reactions, degradation during RNA purification protocols, and variable results in RPAs and Northerns. RNaseZap® contains three ingredients active against RNase and has proven to be extremely effective at removing RNase contamination from glassware, plastic surfaces, countertops, and pipettors. It has also been shown to be effective at eliminating RNase contamination from microfuge tubes without inhibiting subsequent enzymatic reactions. Supplied in one 250 mL bottle.

RNase contamination in microfuge tubes
While concern about RNase contamination of frequently handled labware is common and prudent, researchers often do not worry about RNase contamination in commercially prepared products such as microfuge tubes. However, in a survey of six standard 1.5 mL microfuge tubes in which three were pre-rinsed with RNaseZap® and water and three were left untreated, all of the untreated tubes had at least marginal levels of RNase contamination, whereas none of the tubes pre-rinsed with RNaseZap® exhibited contamination. Additionally, in testing commercially available "RNase-free" microfuge tubes, approximately 10% had some RNase contamination (data not shown). This may well account for the erratic results that researchers periodically experience when performing RNA-based assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: RNase Control Reagent
Form: Liquid
Product Line: Ambion™, RNaseZap®
Product Size: 4 L
Shipping Condition: Room Temperature

Contents & storage


Store at room temperature. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols