Invitrogen™

ElectroZap™ Electrode Decontamination Solution

Catalog number:  AM9785

 Related applications: RNA Extraction | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® ElectroZap™ is specially formulated to instantaneously eliminate RNase contamination from electrodes. Supplied in one bottle containing 250 mL.

• Easily rinsed away leaving no residue
• Stable, safe, and non-corrosive
• No need for strong acid or alkali
• Also removes DNases and other proteins

Researchers performing RNA analysis often require the use of a pH meter to prepare their RNase-free solutions. The use of the same pH meter by several laboratory personnel, however, can often lead to problems with RNase contamination. This is especially a problem when preparing solutions that cannot be DEPC-treated or autoclaved. Special precautions must be taken when removing RNase contamination from a pH meter so that the accuracy will be unaffected and there is no residue left by the cleaning solution. Ambion® ElectroZap™ is a non-corrosive, stable cleaning solution that has been specially formulated to clean electrode surfaces in a simple, quick, and efficient manner. It is effective even against high levels of RNase or DNase contamination. ElectroZap® is simply sprayed directly onto the electrode and then rinsed off with RNase-free water. It does not leave behind any residue that may interfere with subsequent enzymatic reactions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: RNase Control Reagent
Form: Liquid
Product Line: Ambion™, ElectroZap
Product Size: 250 mL
Shipping Condition: Room Temperature

Contents & storage

Store ElectroZap™ at room temperature.

Documents

Manuals & protocols

Recommended products