RNaseZap™ RNase Decontamination Wipes Refill

Catalog number: AM9788

Invitrogen™  Related applications: RNA Extraction | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® RNaseZap Wipes are RNaseZap® in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water. Supplied as 3 rolls of 100 sheets plus solution to refill existing pop-up dispenser.
• Convenient towel format
• Completely removes RNase contamination from glass and plastic surfaces Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free
• Works immediately on contact; no baking or autoclaving necessary The popular Ambion® RNaseZap® is now provided in a convenient wipe format. RNaseZap® Wipes are designed to eliminate RNase contamination on work surfaces, pipettors and equipment that must be RNase-free. RNaseZap® is a formulation of three ingredients known to be active against RNase. It effectively removes even high levels of RNase contamination that similar products cannot. With thorough rinsing, RNaseZap® leaves no residues that are inhibitory to enzymatic reactions. Note: The components are packaged separately. The wipes and RNaseZap® Solution are loaded into the pop-up dispenser prior to first use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: RNase Control Reagent
Form: Wipes
Product Line: Ambion™, RNaseZap®
Product Size: 300 sheets
Shipping Condition: Room Temperature

Contents & storage


Store at room temperature. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products