Invitrogen™

TAE (25X)

Catalog number:  AM9870

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® 25X TAE (High Purity Tris-Acetate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitive application. 10 packets are provided, each containing the necessary powder to make 1 liter of 25X TAE buffer upon the addition of H2O. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Specifications:
Appearance: White crystalline powder Molecular Weight: Tris-Acetate buffer component - 121.14; EDTA component - 372.3
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Running Buffers
Concentrated: 25 X
Gel Compatibility: Agarose Gels
Product Line: Ambion™
Product Size: 10 pack
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols