Related applications: PCR | Real Time PCR (qPCR)
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Reagent Type: | DNA Decontamination Reagent |
---|---|
Form: | Liquid |
Product Line: | Ambion™, DNAZap™ |
Product Size: | 250 mL |
Shipping Condition: | Room Temperature |
Catalog number: | AM10102 |
Size | 1 roll |
Price | USD 129.00 |
Add to favorites | |
Catalog number: | AM1556 |
Size | 40 preps |
Price | USD 362.00 |
Add to favorites | |
Catalog number: | KHO0101 |
Size | 96 tests |
Price | USD 510.00 |
Add to favorites | |