DEPC-Treated Water

Catalog number: AM9922

Ambion™

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® DEPC-treated water is certified nuclease-free and supplied in one bottle containing 1000 mL. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Purity or Quality Grade: DNase-Free, RNase-Free, Molecular Biology Grade
Sample Treatment: DEPC-Treated
Format: Bottle(s)
Product Size: 1 x 1,000 mL
Purification: Membrane-Filtered, Autoclaved
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols