RT-PCR Grade Water

Catalog number:  AM9935

Invitrogen™  Related applications: Real Time PCR (qPCR) | Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

RT-PCR Grade Water is certified free of nucleases, and free of nucleic acid contamination that may cause false-positive signals in RT-PCR. The RT-PCR Grade Water is ready to use and requires no preparation, mixing, or autoclaving.

Quality control
RT-PCR Grade Water is tested for prokaryotic as well as eukaryotic genomic DNA contamination using an ultrasensitive PCR assay. It is provided in 10 x 1.5 mL tubes. Prokaryotic DNA contamination is assessed by performing PCR with primers to 16S rRNA. Eukaryotic DNA contamination is assessed by performing PCR with primers to 18S rRNA. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity and is suitable for any PCR application.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Purity or Quality Grade: DNase-Free, Genomic DNA-Free, Molecular Biology Grade, RNase-Free
Sample Treatment: Not DEPC-Treated
Format: Tube(s)
Product Line: Ambion™
Product Size: 10 x 1.5 mL
Purification: Autoclaved, Membrane-Filtered
Shipping Condition: Room Temperature

Contents & storage

Store at -20°C.

Documents

Manuals & protocols

Recommended products