EVOS™ Onstage Incubator Gas Line
Catalog number: AMEP4732
 Related applications: Cellular Imaging
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The EVOS® Onstage Incubator Gas Line connects the Control Unit and the Stagetop Environmental Chamber units of the EVOS® Onstage Incubator and helps precisely control the CO2 and O2 levels. When performing live cell imaging studies, it is essential that the environmental conditions are precisely controlled. The EVOS® Onstage Incubator has three gas line ports, enabling studies of cells under normal, as well as hypoxic, conditions.
Explore the entire EVOS® line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): EVOS® FL Auto Imaging System
Product Line: EVOS®
Product Size: 1 each
Quantity: 1 each
Type: Onstage Accessory
Documents
Manuals & protocols
Recommended products