EVOS™ Condenser Slider, Blue
Catalog number: AMEPCFB
 Related applications: Cellular Imaging
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Condenser sliders are used to improve illumination under certain conditions and several are available for the EVOS® imaging systems, including a diffuser slider, pinhole slider, and meniscus sliders. Due to variances in sample size, color and thickness, actual slider use may differ from suggestions. This blue slider is intended for brightfield applications at all magnifications as a daylight-correction filter for use with a color camera, like the one included with the EVOS® FL Color Imaging System. The EVOS® XL system enables direct adjustment of the white light LED color temperature.
Explore the entire EVOS® line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): EVOS® FL Auto Imaging System, EVOS® FL Color Imaging System, EVOS® FL Imaging System, EVOS® XL Imaging System
Product Line: EVOS®
Product Size: 1 each
Quantity: 1 each
Type: Light Condenser
Documents
Manuals & protocols
Recommended products