Countess™ II Automated Cell Counter

Catalog number:  AMQAX1000

 Related applications: Cell Counting, Viability, & Cryopreservation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The new Countess™ II Automated Cell Counter is a benchtop assay platform equipped with state-of-the-art optics, full autofocus, and image analysis software for rapid assessment of cells in suspension.

Accurate—use the Countess II Automated Cell Counter to eliminate the subjectivity of manual cell counting and user-to-user variability
Fast—the Countess II Automated Cell Counter provides a total cell count, reports viability, and measures average cell size in as little as 10 seconds
Convenient—the Countess II Automated Cell Counter requires no cleaning or routine maintenance. A capacitive touchscreen and simple user interface provides quick startup and requires minimal training.

Learn more about our automated cell counters >

The Countess II Automated Cell Counter comes with:
• Countess USB drive for storing and transferring data to your computer
• Disposable slide holder
• Power cord and 4 plug adaptors to accommodate most countries
• Quick reference card (QRC)
• A free box of 50 disposable slides

The Countess II Automated Cell Counter works in 3 simple steps:
1. Mix 10 µL of sample with 10 µL of trypan blue and pipet into a disposable Countess chamber slide.
2. Insert the slide into the instrument.
3. Press the "Count" button.

Extended Warranty Packages
Protect your instrument and your investment with Countess II Extended Warranty Packages and save up to 25% off the individual instrument and extended warranty list prices. Learn more >

Get accurate cell counts
The Countess II Automated Cell Counter uses trypan blue staining combined with an auto-focus mechanism and sophisticated image analysis algorithm to obtain accurate cell and viability counts. The measurement range extends from 1 x 10e4 to 1 x 10e7 cells/mL (the optimal range is 1 x 10e5 to 4 x 10e6 cells/mL), which is broader than that of a hemocytometer. The optimal cell size for accurate viability assessment is 7–60 µm, while particles as small as 4 µm can be successfully counted.

Transfer data and images to your PC
Using the USB port on the Countess II Automated Cell Counter, you can save your cell count results and images, and then transfer them to your PC. (For your convenience, a USB drive is supplied with the Countess II instrument).

Countess II/II FL Software >


For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: One 2-chamber disposable slide
Cell Size: 4-60 µm (detection), 7-60 µm (viability)
Concentration: 1x10^4-1x10^7 cells⁄mL
Detection Method: Brightfield
Dimensions: 9 in (228.6 mm) W x 5.5 in (139.7 mm) D x 9 in (228.6 mm) W
Display Interface: 7" Capacitive touch screen
Field: 2.15 mm x 1.62 mm (3.48 mm2)
Input Voltage: 100-240 VAC
Instrument Memory: USB Stick
Product Line: Countess®
Product Size: 1 instrument
Shipping Condition: Room Temperature
Weight: 8 lbs

Documents

Manuals & protocols