Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

BacMam Enhancer Kit

Catalog number: B10107

Invitrogen™  Related applications: Cell Structure | Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The BacMam Enhancer helps to increase expression of BacMam reagents in mammalian cells.

To use, simply add the BacMam enhancer to cell culture media with or without serum that have already been treated with the BacMam reagent, then incubate cells for 1.5–2 hours at 37°C and 5% CO2 and replace with fresh medium. Different cell types vary in their response to the BacMam enhancer and for best results the amount and treatment time will need to be optimized. Once prepared, the enhancer solution is stable to multiple freeze⁄thaw cycles following the recommended storage conditions. If desired, the solution can be divided into aliquots following reconstitution.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Inhibitor Type: Histone Deacetylase Inhibitor
Product Line: Molecular Probes®
Product Size: 1 kit
Specific Inhibitor Target: Histone deacetylase
Shipping Condition: Room Temperature

Contents & storage

Each kit contains sufficient material for treating 10 X 96 microplates. Store at 2—6°C, do not freeze. When stored as directed the product is stable for 6 months.

Documents

Manuals & protocols

Recommended products