Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

BCECF, AM (2',7'-Bis-(2-Carboxyethyl)-5-(and-6)-Carboxyfluorescein, Acetoxymethyl Ester)

Catalog number:  B1150

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

With a pKa of ~6.98, the cell-permeant, dual-excitation ratiometric pH indicator BCECF, AM is ideal for measuring changes in the cytosolic pH of most cells. Intracellular pH measurements with BCECF are made by determining the pH-dependent ratio of emission intensity (detected at 535 nm) when the dye is excited at ~490 nm versus the emission intensity when excited at its isobestic point of ~440 nm.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: pH Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Microplate Reader
Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products