Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
BAPTA, AM, cell permeant chelator
Catalog number:  B1205
Invitrogen™  Related applications: Cell Viability, Proliferation & Function
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
BAPTA, AM is a cell-permeant chelator, which is a highly selective for Ca2+ over Mg2+ , and it can be used to control the level of intracellular Ca2+. BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Indicator Type: Chelator (Non-Fluorescent)
Cell Permeability: Cell-Permeant
Product Size: 25 mg
Shipping Condition: Room Temperature
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols
Recommended products