Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more
Invitrogen™

α-Bungarotoxin, Alexa Fluor™ 594 conjugate

Catalog number:  B13423

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

α-Bungarotoxin, a 74—amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the α-subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions. Our bright and photostable Alexa Fluor 594 α-bungarotoxin may prove to be one of the best probes for visualizing this receptor. Fluorescent α-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and to localize neuromuscular junctions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation⁄Emission (nm): 590⁄617
Product Line: Alexa Fluor™
Product Size: 500 µg
Protein Subtype: Other Proteins
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products