Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

5-(Bromomethyl)Fluorescein

Numéro de catalogue:  B1355

Invitrogen™  Applications liées: Labeling Chemistry | Protein Labeling & Crosslinking

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

5-(bromomethyl)fluorescein can be used to label thiols with the green-fluorescent label, fluorescein. When compared with the iodoacetamide and maleimide derivatives, this thiol-reactive form of fluorescein reacts more slowly, but forms stronger thio-ether bonds.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Chemical Reactivity: Thiol
Label or Dye: FITC (Fluorescein)
Product Size: 10 mg
Shipping Condition: Room Temperature

Contenu et stockage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuels et protocoles

Produits recommandés