Prepare stock solutions with confidence using fresh, single-use size anhydrous DMSO (order D12345)
Invitrogen™
BODIPY™ FL Ouabain
Catalog number:  B23461
 Related applications: Cellular Imaging
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Ouabain is a member of a class of glycosylated steroids collectively known as cardiac glycosides due to their therapeutic efficacy in the treatment of congestive heart failure. Ouabain inhibits the ion transport activity of the plasma membrane Na+/K+-ATPase via binding to its catalytic α-subunit. This BODIPY FL ouabain derivative offers some valuable properties. Because the BODIPY FL fluorophore is nonpolar and relatively small, labeled ligands typically have superior receptor-binding properties over fluorescein- and NBD-labeled derivatives.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line: BODIPY™
Product Size: 100 µg
Shipping Condition: Room Temperature
Contents & storage
Store in freezer (-5 to -30°C) and protect from light.
Documents
Manuals & protocols