Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Biotin-XX, SSE (6-((6-((Biotinoyl)Amino)Hexanoyl)amino)Hexanoic Acid, Sulfosuccinimidyl Ester, Sodium Salt)

Catalog number: B6352

Invitrogen™  Related applications: Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Biotin-XX, SSE reacts efficiently and reproducibly with amines. The biotin is well separated from the point of attachment by a 14-atom spacer consisting of two, seven-atom aminohexanoyl groups (X). This spacer enhances the accessibility of biotin to the relatively deep biotin-binding sites of avidin and streptavidin. The sulfonate group on the succinimidyl ester increases the water solubility of the reactive moiety.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Biotin-XX
Chemical Reactivity: Amine
Product Size: 25 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products