NativePAGE™ Novex™ 3-12% Bis-Tris Protein Gels, 1.0 mm, 10-well

Catalog number: BN1001BOX

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The NativePAGE™ Novex® Bis-Tris Gel System is a pre-cast polyacrylamide mini gel system that provides a sensitive and high-resolution method for analysis of native membrane protein complexes, native soluble proteins, molecular mass estimations, and an assessment of the purity of native proteins. It is based on the blue native polyacrylamide gel electrophoresis technique (BN PAGE) developed by Schagger and von Jagow. With the NativePAGE™ Novex® Bis-Tris Gel System, you will:

• Simplify resolution of large proteins (15–10,000 kDa)
• Analyze membrane-protein complexes in their native conformations
• Obtain better resolution than with traditional Tris-glycine native electrophoresis

Using NativePAGE™ Novex® Bis-Tris Gels
In SDS-PAGE, SDS functions as a charge-shift molecule that denatures proteins by conferring on them a net negative charge and enables proteins to migrate towards the anode. BN PAGE uses Coomassie® G-250 as the charge-shift molecule, which binds to proteins and confers a net negative charge while maintaining the proteins in their nondenatured, native state. The near-neutral pH of the NativePAGE™ Novex® Bis-Tris Gel System provides maximum stability of both the proteins and the gel matrix, resulting in a highly sensitive method for analysis of native membrane-protein complexes and offering superior band resolution over traditional Tris-glycine gel systems.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Percentage: 3-12 %
Wells: 10 wells
Gel Type: Native Gel, Bis-Tris Gel
Separation Range: 30 kDa to 10,000 kDa
Gel Size: Mini (8 cm x 8 cm)
Well Format: 1D Well
Thickness: 1.0 mm
Quantity: 10 gels
Product Size: 10 gels (1 box)
Mode of Separation: Molecular Weight

Contents & storage

One box contains 10 gels. Store at 2°C to 8°C. Do not freeze.

Documents

Manuals & protocols