NativePAGE™ Cathode Buffer Additive (20X)

Catalog number:  BN2002

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NativePAGE™ Cathode Buffer Additive (20X) is added to the NativePAGE™ Running Buffer and the resulting buffer is used in the cathode chamber of an XCell SureLock® Mini Cell for running NativePAGE™ Gels.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Type: IEF Gel
Shipping Condition: Room Temperature
Buffer Type: Anode & Cathode Buffers
Concentrated: 20 X
Gel Compatibility: NativePAGE™ Gels
Product Line: NativePAGE™
Product Size: 250 mL

Contents & storage

Store at room temperature

Documents

Manuals & protocols

Recommended products