New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Digitonin (5%)

Catalog number:  BN2006

 Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

5% Digitonin is a detergent solution used to prepare native protein samples for use with the NativePAGE™ Gel System. Also included in the NativePAGE™ Sample Prep Kit, this ready-to-use detergent solution improves solubility of hydrophobic and membrane proteins during sample preparation. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE™ Sample Prep Kit are compatible with NativePAGE™ Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Size: 1 mL
Reagent Type: Digitonin
Shipping Condition: Wet Ice

Contents & storage

1 ml of solution. Store at +4°C.

Documents

Manuals & protocols

Recommended products