Invitrogen™

NativePAGE™ Sample Prep Kit

Catalog number:  BN2008

 Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The NativePAGE™ Sample Prep Kit includes sample preparation reagents for native gel electrophoresis. The kit includes two ready-to-use detergent solutions, 10% DDM (n-dodecyl-β-D-maltoside) and 5% Digitonin, which improve the solubility of hydrophobic and membrane proteins during sample preparation.

The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE™ Sample Prep Kit are compatible with NativePAGE™ Novex Bis-Tris Gels for native electrophoresis, showing increased resolution and reduced streaking.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Type: Native Gel
Shipping Condition: Wet Ice
Buffer Type: Sample Loading Buffers
Gel Compatibility: NativePAGE™ Gels
Product Line: NativePAGE™
Product Size: 1 kit

Contents & storage

Contains:
• 1 ml of 10% DDM
• 1 ml of 5% Digitonin
• 10 ml of NativePAGE™ 4X Sample Buffer
• 0.5 ml NativePAGE™ 5% G-250 Sample Additive.

Store the kit at 2°C to 8°C.

Documents

Manuals & protocols

Recommended products