Search Thermo Fisher Scientific
Use the Thermo Scientific™ GCI Series Interface to seamlessly integrate your gas chromatography (GC) and inductively coupled plasma mass spectrometry (ICP-MS) systems as an easy-to-use solution for advanced speciation studies. When GC and ICP-MS systems are seamlessly connected, even applications having the most challenging detection limit are no problem. The GCI Series Interface provides easy, reliable measurement of volatile species, down to sub-ppt concentrations.
Type | GC Interface |
Description | GCI 100: GC-ICP-MS Interface for iCAP Q/Qnova |
For Use With (Equipment) | ICP-MS Systems |
Unit Size | Each |
Type | GC Interface |
Description | GCI 200: GC-ICP-MS Interface for ELEMENT 2/XR |
For Use With (Equipment) | ICP-MS Systems |
Unit Size | Each |
Type | GC Interface |
Description | GCI 300: GC-ICP-MS Interface for NEPTUNE Plus |
For Use With (Equipment) | ICP-MS Systems |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Type | Description | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
BRE0008120 | Each | GC Interface | GCI 100: GC-ICP-MS Interface for iCAP Q/Qnova | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BRE0008121 | Each | GC Interface | GCI 200: GC-ICP-MS Interface for ELEMENT 2/XR | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BRE0008122 | Each | GC Interface | GCI 300: GC-ICP-MS Interface for NEPTUNE Plus | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The GCI Series Interface provides: