Gibco™

Human Epidermal Keratinocytes, neonatal (HEKn)

Catalog number:  C0015C

 Related applications: Mammalian Cell Culture | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Primary human epidermal keratinocytes isolated from neonatal foreskin, cryopreserved at the end of the primary culture.
• Viability at least 70%
• ≥500,000 viable cells/vial
• Capable of at least 30 population doublings
• Mycoplasma: Not Detected
• Hepatitis B: Not Detected
• Hepatitis C: Not Detected
• HIV-1: Not Detected
• Bacteria, yeast, and other fungi: Not Detected
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Age: Newborn (14 days or less)
Cell Type: Keratinocytes
Culture Type: Adherent Cell Culture
Donor Source: Single Donor
Form: Cryopreserved
Product Line: Cascade Biologics®
Product Size: 1 vial
Species: Human
Tests Performed: Growth Performance Assay, HIV-1, Hepatitis B (HBV), Hepatitis C (HCV), Mycoplasma, Sterility Testing (Bacteria & Fungi)

Contents & storage

Contents: One vial of cells.

Storage: Liquid nitrogen.

Documents

Manuals & protocols

Recommended products