Human Pulmonary Artery Smooth Muscle Cells (HPASMC), Proliferating Culture, Prepared in Medium 231 and SMGS

Catalog number: C00925PA

Gibco™  Related applications: Mammalian Cell Culture | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Quaternary cultures established from cryopreserved cells, grown to approximately 50% confluence in T-25 flasks.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Age: Adult
Form: Proliferating Culture
Species: Human
Culture Type: Adherent Cell Culture
Donor Source: Pooled
Product Size: 6 x T25 flasks
Tests Performed: Hepatitis B (HBV), Sterility Testing (Bacteria & Fungi), Growth Performance Assay, Mycoplasma, Hepatitis C (HCV), HIV-1
Shipping Condition: Room Temperature

Contents & storage

Upon receipt, follow the instructions in the manual.

Documents

Manuals & protocols