Human Microvascular Endothelial Cells, adult dermis (HMVECad)

Catalog number: C0115C

Gibco™  Related applications: Mammalian Cell Culture | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Primary human microvascular endothelial cells cryopreserved from adult dermis at the end of the tertiary culture.
• Viability at least 70%
• ≥500,000 viable cells/vial
• Capable of at least 16 population doublings
• Mycoplasma: Not Detected
• Hepatitis B: Not Detected
• Hepatitis C: Not Detected
• HIV-1: Not Detected
• Bacteria, yeast, and other fungi: Not Detected
• Positive for von Willebrand factor (vWf)
• Positive for CD31
• Positive for CD36
• Negative for α-actin
• Positive for diI-Ac-LDL uptake
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Cryopreserved
Species: Human
Cell Type: Endothelial Cells (Microvascular)
Donor Source: Single Donor
Age: Adult
Culture Type: Adherent Cell Culture
Product Size: 1 vial
Tests Performed: Hepatitis B (HBV), HIV-1, Sterility Testing (Bacteria & Fungi), Growth Performance Assay, Mycoplasma, Hepatitis C (HCV)

Contents & storage

Store at liquid nitrogen temperature

Documents

Manuals & protocols