Click-iT® TUNEL Alexa Fluor® 488 Imaging Assay, for microscopy & HCS This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: C10245

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function | Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Click-iT® TUNEL Alexa Fluor® 488 Imaging Assay utilizes a dUTP modified with an alkyne, a small, bio-orthogonal functional group that enables the nucleotide to be more readily incorporated by TdT than other modified nucleotides, including BrdUTP, biotin-dUTP or fluorescein-dUTP. Detection is based on a click reaction, a copper (I) catalyzed reaction between an azide and alkyne. Click chemistry fills the void when methods such as direct labeling or the use of antibodies are not efficient. The small size of the detection reagent, an Alexa Fluor® azide (MW <~1000) compared to that of an antibody (MW ~150,000) enables facile penetration of complex samples with only mild fixation and permeabilization required.

Compared to other assays using one of the other modified nucleotides, the Click-iT® TUNEL assay is able to detect a higher percentage of apoptotic cells under identical conditions. The assay is also fast and is complete within 2 hours. Furthermore, the Click-iT® TUNEL assay allows multiplexing with surface and intracellular biomarker detection.

More tools for apoptosis detection and measurement >
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
Format: 96-well plate
Label or Dye: Alexa Fluor™ 488, Hoechst 33342
Product Size: 1 kit
Emission Class: Visible, UV
Detection Method: Fluorescent
Excitation Class: Visible, UV
For Use With (Equipment): Fluorescence Microscope, High Content Instrument
Green Features: Less hazardous
Shipping Condition: Dry Ice

Contents & storage

1 kit containing sufficient reagents to perform 1 X 96 tests or 50 coverslips.
Store at ≤-20°C and protected from light.

Documents

Manuals & protocols

Recommended products