Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Click-IT™ Geranylgeranyl Alcohol, Azide, mixed isomers This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  C10249

 Related applications: Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Identify and characterize geranylated proteins with Click-iT® geranylgeranyl alcohol, azide using the powerful click chemistry, a simple and robust two-step labeling and detection technique. In step one, the azide-containing biomolecule is fed to cells or animals and actively incorporated into proteins. Unlike other labels such as biotin or a fluorescent dye, the azide-tag is small enough that the tagged molecule is an acceptable substrate for the enzymes that incorporate this building block into proteins. Detection utilizes the chemoselective ligation or “click" reaction between and azide and an alkyne where the modified protein is detected with the corresponding alkyne-containing dye or hapten using either the Click-iT® Cell Reaction Buffer Kit or the Click-iT® Protein Buffer Kit. With the Click-iT® Cell Reaction Buffer Kit, cells can be analyzed by fluorescence microscopy, flow cytometry or high-content imaging and analysis (HCS) together with other biomarkers of interest for content and context rich results. With the Click-iT® Protein Reaction Buffer Kit, achieve detection sensitivity in 1-D gels and western blots in the low femtomole range or perform LC-MS⁄MS and MALDI MS analysis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Solid
Green Features: Less hazardous
Label or Dye: Azide
Labeling Method: Metabolic Labeling
Labeling Target: Proteins (General), Proteins (Nascent)
Product Line: Click-iT™, Molecular Probes®
Product Size: 1 mg
Shipping Condition: Dry Ice

Contents & storage

Store at ≤-20°C, desiccated and protected from light.

Documents

Manuals & protocols

Recommended products