Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Click-iT™ Protein Reaction Buffer Kit This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  C10276

 Related applications: Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Click-iT® Protein Buffer Kit provides researchers who have proteins labeled with an azide or alkyne and the corresponding click detection reagent with the means to click these two moieties together. Each kit includes sufficient material to perform 25 reactions for subsequent analysis by gel electrophoresis, western blot or mass spectrometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Number of Reactions: 25 Reactions
Product Line: Click-iT™, Molecular Probes®
Product Size: 1 kit
Green Features: Less hazardous
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products