Click-iT® RNA Alexa Fluor® 488 HCS Assay This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Click-iT® RNA Alexa Fluor® 488 HCS Assay enables detection of global RNA transcription temporally and spatially in cells (PNAS (2008) 105:15779-84). The ability to detect newly synthesized RNA or changes in RNA levels resulting from disease, environmental damage or drug treatments is an important aspect of toxicological profiling. Utilizing an alkyne-modified nucleoside, 5-ethynyl uridine (EU), and powerful click chemistry, newly synthesized RNA can be detected without the use of radioactivity or antibodies with a simple, two-step procedure. In step one, the alkyne-containing nucleoside is fed to cells or animals and actively incorporated into nascent RNA. The small size of the tag enables efficient incorporation of the modified nucleoside into RNA, but not into DNA. Detection utilizes the chemoselective ligation or “click" reaction between and azide and an alkyne where the modified RNA is detected with a corresponding azide-containing dye. With its dimunitive “footprint", the Click-iT® detection molecule can easily penetrate complex samples and leaves open the possibility of multiplex analyses with other probes, including antibodies for the detection of RNA-interactive proteins for deeper biological insights.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
Detection Method: Fluorescent
Emission Class: Visible
Excitation Class: Visible
Excitation⁄Emission (nm): 495⁄519
For Use With (Equipment): High Content Instrument
Format: 96-well plate
Label or Dye: Alexa Fluor™ 488
Product Line: Alexa Fluor®, Click-iT™, Molecular Probes®
Product Size: 1 kit
Green Features: Less hazardous
Shipping Condition: Room Temperature

Contents & storage

The kit contains sufficient material for 2 X 96-well plates. Store at 2-6°C, dessicate and protect from light.

Documents

Manuals & protocols

Recommended products