All the benefits of Click-iT® EdU—now with expanded multiplex capabilities. Click here
Proliferating cells revealed: Click-iT™ EdU Colorimetric assays for IHC

Click-iT® EdU Alexa Fluor® 647 Imaging Kit This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: C10340

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Click-iT® EdU Alexa Fluor® 647 Imaging Kit is a superior alternative to traditional proliferation assays that is optimized for fluorescence microscopy applications. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor® dye in a fast, highly-specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol.

• Simple—works the first time, every time, in less time
• Efficient—no denaturation steps or harsh treatment required
• Content-rich results—better preservation of cell morphology, antigen structure, and DNA integrity
• Consistent—not dependent on variable antibody lots for detection

The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently. The harsh treatment can also adversely effect sample integrity and quality, which makes co-staining with other antibodies challenging.

Superior Proliferation Methodology
The Click-iT® EdU Alexa Fluor® 647 Imaging Kit provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT® EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT® EdU detection reagent to gain access to the DNA. As a consequence, the Click-iT® EdU imaging kit is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.

This kit is optimized for fluorescence microscopy applications; visit the Click-iT® technology area of our website for kits designed for high-content imaging, fluorescence microplate, or flow cytometry platforms.

Learn more about Click-iT technology >
Find more tools for image-based detection of proliferating cells >

Notes:
The Click-iT® assay can be used on cells in culture or in vivo following EdU administration by feeding or injection methods.
The Click-iT® assay can be used with BrdU in dual pulse experiments by using the anti-BrdU (clone MoBu-1) antibody, which does not cross react with EdU.
The Click-iT® technology is compatible with immunohistochemical, immunocytochemical, and fluorescent dyes that are fixation tolerant or designed for fixed cell labeling.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Confocal Microscope, Fluorescence Microscope
Detection Method: Fluorescent
Format: Slide(s)
Color: Far-Red
Emission Class: near IR
Excitation Class: near IR
Excitation⁄Emission (nm): 650⁄668
Label or Dye: Alexa Fluor™ 647
Product Line: Alexa Fluor®, Click-iT™, Molecular Probes®
Product Size: 1 kit
Green Features: Less hazardous
Shipping Condition: Room Temperature

Contents & storage

The kit contains sufficient material for 50 18x18 coverslips. Store at 2-6°C, dessicate and protect from light.

Documents

Manuals & protocols

Recommended products