Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

Click-IT™ Alexa Fluor™ 594 DIBO Alkyne, for copper free click chemistry detection of azide This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  C10407

 Related applications: Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The fluorescent Alexa Fluor® 594 Dibo alkyne is reactive with azides via a copper-free "click chemistry" reaction.

Please consider this copper-free variation to our copper requiring alkynes if you are staining the surface of live cells or have concerns about native protein function loss with copper in cell extracts. Copper can damage fluorescent proteins, Quantum Dot nanocyrstals, certain enzymes and photoproteins like RPE. The DIBO reagent is not suitable for staining intracellular components of fixed and permeabilized cells due to high backgrounds.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Red-Orange
Detection Location: In-Cell Detection
Detection Method: Fluorescent
Emission Class: Visible
Excitation Class: Visible
Excitation⁄Emission (nm): 594⁄617
For Use With (Equipment): Confocal Microscope, Flow Cytometer, Fluorescence Microscope, High Content Instrument
Form: Solid
Green Features: Less hazardous
Label or Dye: Alexa Fluor™ 594
Product Line: Click-iT™
Product Size: 500 µg
Shipping Condition: Dry Ice
Solubility: DMSO (Dimethylsulfoxide)

Contents & storage

Store at ≤ -20°C. Desiccation recommended. Protect material from long-term exposure to light; may be exposed to light for short periods of time

Documents

Manuals & protocols