Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.

Click-iT™ Protein Enrichment Kit, for click chemistry capture of azide-modified proteins

Catalog number:  C10416

Invitrogen™  Related applications: Labeling Chemistry | Protein Mass Spectrometry Analysis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The Click-iT® Protein Enrichment Kit for the efficient capture of click chemistry based azide-modified proteins on a resin of alkyne agarose. Superior to biotin or lectin based enrichment approaches. Ideal for proteomics, biomarker discovery, posttranslational modification (PTM) analysis, and more. The azide modification can occur via metabolic feeding, enzymatic addition, or chemical modification. Click-Azide modified proteins, or their post-translationally modified forms, are enriched from complex protein extracts on the alkyne-agarose resin supplied. Once anchored to the resin via copper catalyzed click chemistry, extensive washing yields a highly enriched population of nascent molecules that can be furthered characterized by mass spectrometry. The alkyne-agarose resin improves upon existing biotin approaches by>8 fold, with signal to noise of biotin = 3 while the Click-iT resin = 25.

Perfect upstream MS-based sample preparation technique for looking at changes in global protein expression and biomarker analysis.

The new Click-iT® enrichment resin affords many distinct advantages including::

  1. global differential profiling of multiple subclasses of posttranslationally modified (PTM) proteins and newly synthesized proteins

  2. improved signal to noise by eliminating nonspecific binding and increasing selectivity, thus improving detection of low abundant proteins

  3. accelerates PTM site identification leading to more rapid initiation of functional studies

  4. fully compatible with widely used MS techniques including iTRAC and ICAT.

Seamless integration of cell biology with protein chemistry.

Affordable, easy, no special equipment required.

To know what’s new in your cells use Click-iT®

Click-iT®: One Reaction, Endless Possibilities.
For Research Use Only. Not for use in diagnostic procedures.


Format: Spin Column
Isolation Technology: Click Chemistry
Product Line: Click-iT™
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store at 2-8°C. Desiccation recommended. Protect material from long-term exposure to light; may be exposed to light for short periods of time


Manuals & protocols