Invitrogen™

CellROX™ Green Flow Cytometry Assay Kit

Catalog number:  C10492

 Related applications: Cell Viability, Proliferation & Function | Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CellROX™ Green Flow Cytometry Assay Kit enables the flow cytometric detection of reactive oxygen species (ROS) in live cells. The kit includes the novel, fluorogenic CellROX™ Green Reagent, as well as SYTOX™ Red Dead Cell Stain, N-acetyl cysteine (an antioxidant, for negative control), and tert-butyl hydroperoxide solution (TBHP, an inducer of ROS).

View a selection guide for all CellROX® Reagents and Kits.

The CellROX™ Green Flow Cytometry Assay Kit features:

• Fluorogenic probe formulated for flow cytometry that is oxidized in the presence of ROS
• Multicolor compatibility—minimal overlap with fluorophores excited by other laser lines, allowing easy multiplexing with other reagents
• Simple protocol—cells can be stained in complete media or other appropriate buffer, no need for serum free media

The CellROX™ Green Detection Reagent is cell-permeable and non-fluorescent or very weakly fluorescent while in the reduced state. Upon oxidation, the reagent binds DNA and exhibits a strong fluorogenic signal that has absorption/emission maxima of 508/525 nm and remains localized to the nucleus and cytoplasm. When used together with the included SYTOX™ Red Dead Cell Stain, oxidatively stressed and nonstressed cells are reliably distinguished from dead cells by flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Compatible Cells: Eukaryotic Cells, Mammalian Cells
For Use With (Equipment): Flow Cytometer
Detection Method: Fluorescent
Format: Tube(s)
Excitation⁄Emission (nm): CellROX® green: 508⁄525, SYTOX® red: 640⁄658
Flow Cytometer Laser Lines: 488, 633/635
Label or Dye: CellROX® Green Reagent
Number of Reactions: 100
Product Line: CellROX®
Product Size: 100 assays
Solubility: DMSO (Dimethylsulfoxide)
Shipping Condition: Wet Ice

Contents & storage

Contains 1 vial CellROX® Green Reagent (25 µL), 1 vial SYTOX® red dead cell stain (100 µL), 2 vials N-acetylcysteine (10 mg per vial), 1 vial tert-butyl hydroperoxide (50 µL, 70% solution in water), and 1 vial DMSO (200 µL).

Store kit at -5 to -30°C, protected from light. CellROX® reagent is air sensitive.

Documents

Manuals & protocols