Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

CellLight™ Actin-GFP, BacMam 2.0

Catalog number: C10506

Invitrogen™  Related applications: Cell Structure | Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CellLight® Actin-GFP, BacMam 2.0, provides an easy method for the labeling of actin with Green Fluorescent Protein (GFP) in live cells. Simply add the reagent to your cells, incubate overnight, and the cells are ready to image in the morning. This ready-to-use construct is transduced/transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to human actin. You can observe actin-GFP behavior in live cells using a fluorescent imaging system and standard FITC/GFP filter set. Study dynamic action of actin filaments in live cells, with almost no cytotoxicity. In addition, you can co-transduce/transfect more than one CellLight® reagent and label live cells with multiple tracking or tracing dyes to image dynamic cellular processes. Cells expressing CellLight® constructs can also be fixed with formaldehyde for multiplexing with antibodies using immunocytochemical techniques.

CellLight® Technology
Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
Fast and convenient: simply add CellLight® reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
Less toxic: CellLight® reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling
Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose

Learn more about CellLight® fluorescent protein labeling

BacMam Technology
CellLight® Actin-GFP, BacMam 2.0, is a fusion construct of human actin and emGFP, providing accurate and specific targeting to cellular actin-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight® reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
Delivery Method: Baculovirus (BacMam System)
Detection Method: Fluorescent
Emission Class: Visible
Excitation Class: Visible
Excitation⁄Emission (nm): 488⁄510
Flow Cytometer Laser Lines: 488
For Use With (Equipment): Attune® Acoustic Focusing Cytometer, Confocal Microscope, Floid® Cell Imaging System, Fluorescence Microscope, Microscope
Form: Liquid
Label or Dye: GFP (EmGFP)
Product Line: CellLight®
Product Size: 100 µL
Sub-Cellular Localization: Actin
Technique: Fluorescence Intensity, In Vivo Imaging
Shipping Condition: Wet Ice

Contents & storage

Store at 2-6° C, protected from light. Do Not Freeze.

Documents

Manuals & protocols

Recommended products