Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

Click-iT™ Plus Alexa Fluor™ 647 Picolyl Azide Toolkit

Catalog number:  C10643

 Related applications: Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Click-iT® Plus Alexa Fluor® picolyl azide toolkits contain everything you need to perform copper-catalyzed click reactions with compounds that are sensitive to copper, while retaining all of the benefits of the standard azide/alkyne click reaction. Picolyl azides react efficiently with chelated copper, so free copper in the click reaction can be minimized, protecting against undesired copper side reactions with proteins (e.g., GFP, RPE), nucleic acids (e.g., RNA, oligos), and even small molecules (e.g., phalloidin).

• Flexible—optimize your own Click–iT® reaction for the detection or labeling of any alkyne-containing biomolecule in vitro or in cells and tissue samples
• Bright and sensitive—superior fluorescent Alexa Flour® dyes combined with the efficiency of picolyl azide
• Convenient—contains all the buffers and reagents needed to create the optimal click reaction

Each Click-iT® Plus Picolyl Azide Toolkit includes 100 µg of Alexa Fluor® dye-picolyl azide and all of the reagents required to create a reaction cocktail with the optimal copper and chelate concentration for any click chemistry-based labeling or detection application. Guidelines for designing and optimizing the picolyl azide/alkyne click reaction of your choice are provided in the product manual. Additionally Click-iT® Plus picolyl azides offer increased sensitivity and faster reaction times compared to standard azides using identical reaction conditions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Click-iT™
Product Size: 1 kit

Contents & storage

• 1 x 100 µg Alexa Fluor® dye-picolyl azide
• 1 x 4 mL Click-iT® reaction buffer
• 1 x 1.5 mL copper (II) sulfate
• 1 x 500 µL copper protectant
• 1 Click-iT® buffer additive

Each kit contains enough material for approximately 25 to 100 reactions, depending on application.

Store at 2–8°C. Dessicate and protect from light.

Documents

Manuals & protocols