Cell Sorting Set-up Beads (for UV lasers)

Catalog number:  C16506

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Molecular Probes® Cell Sorting Set-up Beads are reliable standards for flow cytometry sorter instrument setup and calibration. The Cell Sorting Set-up Beads approximate the size, emission wavelength, and intensity of many biological samples, and so can be used to calibrate a flow cytometer’s cell sorting system, including laser source, optics, and stream flow. The beads are optimized for checking cell sorter settings such as drop delay and efficiency (cell loss during sorting).

View information for all flow cytometry calibration and size reference beads.

Cell Sorting Set-up Beads are available for use with UV, blue, green/yellow, and red lasers.These 6.0 µm beads for UV lasers are fluorescent-dye infused microspheres that are excitable between 350–370 nm with emission in the range of 400–470 nm.

Cell Sorting Set-Up Beads provide:

• Consistency in production, creating ideal reference standards
• Confidence through reliable, optimal instrument performance
• Compatibility—use with any instrument

Bead Labeling
As with all of our flow cytometry standard microspheres, the Cell Sorting Set-up Beads are stained internally rather than on the surface. The dyes are therefore insulated from environmental interactions that could cause variable fluorescence output, resulting in excellent signal stability. These 6.0 µm fluorescent polystyrene microspheres are supplied as suspensions packaged in dropper vials for convenient dispensation and are available in a choice of four colors (UV, blue, green/yellow, red) to match your laser.

The dyes used in the manufacture of the Cell Sorting Set-up Beads have been carefully selected to provide emission peaks coincident with cells labeled with commonly used fluorescent dyes. The emission profiles for these standards are intentionally narrow in comparison to cells labeled with a corresponding fluorescent dye. Due to their narrow emission profiles, beads of two different fluorescent colors exhibit minimal spectral overlap, and little or no color compensation is needed when setting up for multicolor experiments.

These beads are an alternative to PeakFlow™ flow cytometry reference beads.

For optimal verification of the laser alignment of a cell sorter cytometer, please see our selection of Alignflow™ Flow Cytometry Alignment Beads.
For Research Use Only. Not for use in diagnostic procedures.


Bead Diameter: 6 µm
Flow Cytometer Laser Lines: UV
Stain Configuration: Core
Calibration Type: Flow Cytometry Reference Beads
Color: Ultraviolet
Detection Method: Fluorescent
Diameter (Metric): 6 µm
Excitation Class: UV
For Use With (Equipment): Flow Cytometer
Form: Suspension
Format: Tube(s)
Product Size: 3 mL
Shipping Condition: Room Temperature

Contents & storage

Contains 2 vials Cell Sorting Set-up Beads (1.5 mL per vial).

Store in refrigerator (2–8°C) and protect from light.


Manuals & protocols

Recommended products