Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

CoverWell™ Incubation Chamber Gasket

Catalog number: C18150

Invitrogen™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CoverWell incubation chamber gaskets are expressly designed for immunocytochemistry and in situ hybridization. The chambers improve the uniformity and sensitivity of staining by enclosing a large sample area while minimizing the reagent volume required. The watertight seal holds reactants in place and prevents evaporation even during prolonged heating and is easily removed for multiple-step procedures. These chamber gaskets are heat resistant, autoclavable and nuclease free.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Depth: 0.2 mm
Dimensions: 40 mm x 22 mm
Product Line: CoverWell™
Product Size: 25 gaskets
Shipping Condition: Room Temperature
Type: Gaskets

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products