Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

CMNB-Caged Carboxyfluorescein, SE (5-Carboxyfluorescein-Bis-(5-Carboxymethoxy-2-Nitrobenzyl) Ether, β-Alanine-Carboxamide, Succinimidyl Ester)

Catalog number: C20050

Invitrogen™  Related applications: Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Conjugation of the succinimidyl ester of the water-soluble, CMNB-caged carboxyfluorescein, SE to a primary amine on a biomolecule of interest produces an essentially nonfluorescent probe that yields a green-fluorescent fluorescein-labeled product only after ultraviolet illumination and removal of the caging group.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: FITC (Fluorescein)
Chemical Reactivity: Amine
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products